Learn More
10L Toluene AR
Brand: Fisher Chemical T/2300/24RSS
106.40 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
10L Toluene AR
Code : 02
Additional Details : CAS Number : 108-88-3 Weight : 8.67000kg Transport : UN number : 1294 Chem class : 3 Pack group : II
Safety and Handling
- Toluene
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.