missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leica Microsystems 10X/18 Eyepiece, DM100/300
Suitable eyepiece for the DM100/300 series of innovative educational compound microscopes. 10X/18 Eyepiece, DM100/300 is available with a reticule or pointer.
£47.50 - £224.00
Specifications
Type | Eyepiece |
---|---|
Field of View | 18 |
For Use With (Equipment) | DM100/300 Series Microscopes |
Magnification Power | 10X |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
12817905
|
Leica Microsystems
13613333 |
Reticule |
£224.00
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
12897895
|
Leica Microsystems
13613331 |
Without pointer or reticule |
£47.50
Each |
Estimated Shipment: 23-01-2025
Log in to see stock. |
|||||
12807905
|
Leica Microsystems
13613332 |
Pointer |
£62.60
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
Description
- For DM100/300 series of educational compound microscopes.
- Available with a reticule or pointer.
Specifications
Eyepiece | |
18 | |
DM100/300 Series Microscopes | |
10X |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title