missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango Buffer (10X)

Product Code. 11541505 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 x 1.0 mL
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 11541505

Brand: Thermo Scientific™ BY5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ensure the optimum reaction conditions for restriction enzymes with Thermo Scientific™ 10X Buffer Tango, premixed with BSA for enhanced stability.

Our Five Buffer System ensures the optimum reaction conditions for each restriction enzyme. This system consists of 10X B (blue), G (green), O (orange), R (red), and Tango (yellow) buffers. All restriction enzymes are supplied in color-coded tubes to indicate the recommended reaction buffer. The recommended buffer and/or the universal Tango buffer are supplied with each enzyme.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

Restriction enzymes exhibit 100% of their certified activity in the recommended buffer. However, some enzymes require additives to achieve 100% activity. For example, AjuI, AlfI, BdaI, BplI, BseMII, FaqI, Eco57I, Eco57MI, Hin4I, and TsoI require S-adenosylmethionine, which is supplied with the enzyme, while AarI and BveI require an oligonucleotide (also supplied with the enzyme). Esp3I requires DTT.

Tango buffer has been designed for double digestions of DNA with conventional restriction enzymes.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 10X
Quantity 5 x 1.0 mL
Product Type Tango Buffer
Research Category Traditional Cloning
Product Line Tango

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.