Learn More
11-Bromo-1-undecene, 90+%, Thermo Scientific Chemicals
£40.70 - £126.00
Chemical Identifiers
CAS | 7766-50-9 |
---|---|
Molecular Formula | C11H21Br |
Molecular Weight (g/mol) | 233.193 |
MDL Number | MFCD00040825 |
InChI Key | YPLVPFUSXYSHJD-UHFFFAOYSA-N |
Synonym | 11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide |
PubChem CID | 284148 |
IUPAC Name | 11-bromoundec-1-ene |
SMILES | C=CCCCCCCCCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484897
|
Thermo Scientific Alfa Aesar
B21849.06 |
5 g |
£40.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494897
|
Thermo Scientific Alfa Aesar
B21849.14 |
25 g |
£126.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
11-Bromo-1-undecene is used in advanced polymer dielectrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications11-Bromo-1-undecene is used in advanced polymer dielectrics.
Solubility
Soluble in water, 0.2422 mg/L @ 25°C (est).
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Chemical Identifiers
7766-50-9 | |
233.193 | |
YPLVPFUSXYSHJD-UHFFFAOYSA-N | |
284148 | |
C=CCCCCCCCCCBr |
C11H21Br | |
MFCD00040825 | |
11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide | |
11-bromoundec-1-ene |
Specifications
7766-50-9 | |
149°C to 150°C (35 mmHg) | |
0.96 | |
1.469 | |
1753231 | |
Soluble in water,0.2422mg/L @ 25°C (est). | |
C=CCCCCCCCCCBr | |
233.193 | |
233.2 | |
11-Bromo-1-undecene |
1.063 | |
>110°C (230°F) | |
C11H21Br | |
MFCD00040825 | |
11-bromo-1-undecene, 10-undecenyl bromide, 1-bromo-10-undecene, undec-10-enyl bromide, 11-bromo-undec-1-ene, 1-undecene, 11-bromo, 11-bromoundecene, undecylenyl bromide, n-undecylenic bromide, w-undecylenyl bromide | |
YPLVPFUSXYSHJD-UHFFFAOYSA-N | |
11-bromoundec-1-ene | |
284148 | |
96% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only