missing translation for 'onlineSavingsMsg'
Learn More

11-Hydroxyundecylphosphonic acid, 95%

Product Code. 15416497
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15416497

Brand: Thermo Scientific Alfa Aesar H33647.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 83905-98-0 | C11H25O4P | 252.291 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83905-98-0
Molecular Formula C11H25O4P
Molecular Weight (g/mol) 252.291
MDL Number MFCD11982869
InChI Key PPCDEFQVKBXBPS-UHFFFAOYSA-N
Synonym 11-hydroxyundecyl phosphonic acid, 11-hydroxyundecyl-1-phosphonic acid, 11-hydroxyundecylphosphonic acid, phosphonic acid, 11-hydroxyundecyl, 11-hydroxyundecylphosphonic acid gc
PubChem CID 45158971
IUPAC Name 11-hydroxyundecylphosphonic acid
SMILES C(CCCCCO)CCCCCP(=O)(O)O

Specifications

Melting Point 110°C to 112°C
Linear Formula HO(CH2)11P(O)(OH)2
Quantity 250 mg
Formula Weight 252.29
Percent Purity 95%
Chemical Name or Material 11-Hydroxyundecylphosphonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.