Learn More
Hanna Instruments™ Chemical Test Kit Replacement Reagents
Pre-made reagents prepared with high-purity chemicals for ease of use. Hanna Instruments™ Chemical Test Kit Replacement Reagents are pre-measured for fast and accurate titrations using the appropriate Hanna Chemical Test Kit.
Brand:  Hanna Instruments™ HI3812-100
 View more versions of this product
Product Code. 11612360
  • £61.20 / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These high quality reagents are manufactured in Hanna's state-of-the-art facility and are clearly marked with the lot number and expiration date on each packet/bottle for traceability. Replacement titration reagent sets provide significant cost savings compared to purchasing an entirely new Test Kit.
Chemical Test Kit Replacement Reagents are available for the following Chemical Test Kits:
  • Acidity Test Kit
  • Alkalinity Test Kit
  • Ascorbic Acid Test Kit
  • Cardon Dioxide Test Kit
  • Chloride Test Kit
  • Dissolved Oxgen Test Kit
  • Free and Total Chlorine (Low, Medium, High Range) Test Kit
  • Free Chlorine Test Kit
  • Total Chlorine Test Kit
  • Iron (Medium Range) Test Kit
  • Phosphate Test Kit
  • Salinity Test Kit
  • Sulfite Test Kit
  • Total Hardness Test Kit
Specifications
Specifications
EDTA titration
HI 3812 Total Hardness Chemical Test Kit
100
Total hardness: 0.0 to 30.0mg/L and CaCO3: 0 to 300mg/L
Reagent Set
Total hardness determination
Bottles with Protective Cap and Instructions
1 Set
Titration