missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- 11 mm Crimp Neck Vial

Specifications
Microliter | |
11.6 mm | |
1.8 mL | |
Crimp | |
Inner Cone | |
1.5 mL | |
32 mm | |
ND11 |
Clear | |
Glass | |
GC, HPLC | |
CleanPack, Polypropylene Box | |
<8 μL | |
Flat Bottom | |
100 | |
No |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Fisherbrand Focus on glassware and plasticware
File size: 66.4 MB
Special Offers
-
Multiple Manufacturers
Incredible Savings on Peptide and Protein Mass Spectrometry Testing Products
Explore our new Peptide and Protein Mass Spectrometry Testing Products PromotionPromo Code: 24142Expires: 2025-03-14
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
16Hours
10Minutes
0Seconds
11 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
46Hours
10Minutes
0Seconds
11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ 11 mm Crimp Neck Glass Microliter Vial > Microliterflat bottom,inner cone,1.1ml,32mm height
Spot an opportunity for improvement?Share a Content Correction