missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ 12 Column Reagent Reservoir with Pyramidal Base
Reagent reservoir for use in automation.
Brand: BRAND™ 701442
196.44 GBP valid until 2024-12-31
Use promo code "22009" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with Liquid Handling Station pipetting robot
- BIO-CERT™ Certified Quality: Free of DNA, RNase, DNase, and pyrogens
Specifications
Reservoir, PP, 12-columns, 12 mL, pyramidal bottom, BIO-CERT™ CERTIFIED QUALITY | |
Polypropylene | |
Reagent Reservoir |
12 mL | |
Non-sterile | |
12 |