missing translation for 'onlineSavingsMsg'
Learn More

12-Tungstophosphate hydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11361868
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

12-Tungstophosphate hydrate is used in histology as a component for staining of cell specimens. It binds to fibrin, collagen and fibers of connective tissues. It is also used to replace the anions of dyes from these materials. In solution, it acts as a homogeneous catalyst and involved in the hydrolysis of propene to give 2-propanol. It is supported with alumina and silica and act as a heterogeneous catalyst involved in the dehydration of 2-propanol to propene and methanol to hydrocarbons. In analytical chemistry, it is used to precipitate proteins and termed as universal precipitant for polar proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12501-23-4
Molecular Formula H5O41PW12
Molecular Weight (g/mol) 2880.20
MDL Number MFCD00011342
InChI Key AVFBYUADVDVJQL-UHFFFAOYSA-N
Synonym phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
PubChem CID 16212977
SMILES O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

Specifications

Melting Point 95°C
Color White through Yellow
Physical Form Powder
Quantity 50 g
UN Number UN3260
Solubility Information Soluble in water,dichloromethane,acetonitrile,acetic acid and ionic liquids.
IUPAC Name phosphoric acid dodecakis(trioxotungsten) hydrate
Formula Weight 2880.20 (Anhydrous)
Grade Reagent
Sensitivity Moisture & Light Sensitive
Chemical Name or Material 12-Tungstophosphate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'