missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow Pumps 120 Series Drive with Pumphead

Product Code. p-200000118
Click to view available options
Motor Speed:
0.1 to 100 rpm
0.1 to 200 rpm
0.1 to 32 rpm
10 rpm
17 rpm
220 rpm
31 rpm
52 rpm
Pump Head:
102R
114DV
400D1
400DM2
400DM3
Model:
120F
120S
120U
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17853084

Brand: Watson-Marlow Pumps 010.6111.DA0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

100 Series Pumps are small and stackable, saving valuable space in cleanroom, LAF and biosafety cabinet installations, capable of flow rates up to 190 mL/min.

101 series pumps are small and stackable lab pumps for handling valuable and delicate fluids without contamination. Save valuable space in cleanrooms, LAF and biosafety cabinets.

  • The 100 series is an ideal pump for maximizing yield in fermentation and cell culture applications.
  • Ultra-compact and stackable for multiple feeds such as pH control, antifoam, nutrient and buffer addition, the 120 offers the ideal combination of size and technology to optimize your process.
  • The 100 series pump is suited to single-use systems.
  • Error-free tube change takes only seconds.
  • Biopharmaceutical processes require accurate and repeatable pumping that is easy to validate.
  • With a 2,000:1 speed range, the 100 series pump delivers the flexibility and purity so vital to drug trials.
  • A keypad lock secures your pumping parameters for consistent and repeatable performance.
Pumphead options available as standard (others available on request):
  • 120R single channel, two roller pumphead for continuous tubing
  • 114DV flip-top single-channel pumphead for continuous tubing
  • 400D1 single-channel pumphead for continuous tubing
  • 400DM2 twin-channel pumphead for manifold tubing
  • 400DM3 three-channel pumphead for manifold tubing
TRUSTED_SUSTAINABILITY

Specifications

Control Type Analog
Display Type LED
Enclosure Rating IP31
Flow Rate 0.002 to 190 mL/min.
Maximum Pressure 30 psi
Motor Speed 10 rpm
Model 120F
Pump Head 114DV
Voltage 90/264 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.