missing translation for 'onlineSavingsMsg'
Learn More

Westfield Medical Sterilisation pouch gussetted, chevron shaped end with thumb holes 200mm x 50mm x 400mm

Brand:  Westfield Medical WPG200X400

Sterilisation pouch gussetted, chevron shaped end with thumb holes 200mm x 50mm x 400mm

Code : 03

Additional Details : Weight : 0.20000kg

Product Code. 12749035

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers