missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfur Set for ASTM D5453: S @ 0, 2.5, 5, 7.5, 15, 20 ng/μL in p-xylene, VHG™
Click to view available options
Quantity:
6 x 2 mL
Unit Size:
12mL
Description
Sulfur Set for ASTM D5453: S @ 0, 2.5, 5, 7.5, 15, 20 ng/μL in p-xylene
Specifications
Specifications
| Chemical Name or Material | Sulfur |
| CAS | 7704-34-9 |
| Analyte or Component Names | Sulfur |
| Shipping Condition | Room Temperature |
| Molecular Formula | S |
| InChI Formula | InChI=1S/S |
| SMILES | [S] |
| IUPAC Name | sulfur |
| Solvent or Matrix | p-xylene |
| Molecular Weight (g/mol) | 32.065 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction