Learn More
Sartorius™ 13101 Series Non-sterile Cellulose Nitrate (CN) Membrane Filters
Brand: Sartorius™ 13101--50----AHN
Code : NEW
Additional Details : Weight : 0.05000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Specifications
White | |
Cellulose Nitrate | |
8 μm | |
100 Pack | |
Membrane Filter | |
50 mm |
Membrane Filter | |
Cellulose Nitrate | |
Non-sterile | |
115 to 145 μm | |
50 mm |
Safety and Handling
- MEMBRANE NC EDGE W N8µ 50mm
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose