missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 13mm Crimp Cap

Product Code. 12636685 Shop All Fisherbrand Products
Click to view available options
Color:
Clear Lacquered
Unit Size:
Pack of 1000

Product Code. 12636685

Only null left
Add to basket
Add to basket

Provide a secure leak-resistant seal.

  • 13mm
  • aluminum Cap
  • Cclear lacquered
  • Center hole
  • Aluminum seals must be applied with a crimping tool
TRUSTED_SUSTAINABILITY

Specifications

Color Clear Lacquered
Diameter (Metric) Outer 13 mm
Material Aluminum
Quantity 100
Type Crimp Cap
Closure Size ND13
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.