missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 beta/zeta Monoclonal Antibody (4E2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125665
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa cell lysate.
14-3-3 beta/zeta Monoclonal antibody specifically detects 14-3-3 beta/zeta in Bovine, Canine, Human, Mouse, Non-human Primate, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
14-3-3 beta/zeta | |
Monoclonal | |
1 mg/mL | |
PBS with 0.1mM PMSF, 50% glycerol and 0.09% Sodium azide; pH 7.2 | |
P63103, P68250, P31946, P63104, P63101, Q9CQV8, P35213, P63102 | |
YWHAB, YWHAZ | |
Lysed, transformed, human amnion cells. | |
50 μg | |
Primary | |
Bovine, Canine, Human, Mouse, Non-human Primate, Porcine, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
4E2 | |
Unconjugated | |
YWHAB | |
1110013I11Rik; 1300003C17Rik; 14-3-3 alpha; 14-3-3 delta; 14-3-3 protein beta; 14-3-3 protein beta/alpha; 14-3-3 protein beta/alpha, N-terminally processed; 14-3-3 protein beta-subtype; 14-3-3 protein zeta; 14-3-3 protein zeta/delta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 zeta; 14-3-3d; 14-3-3z; 14-3-3zeta; 14-3-3-zeta; 3-monooxygenase/tryptophan 5 monooxygenase activation protein beta polypeptide; AI596267; AL022924; AU020854; brain protein 14-3-3, beta isoform; epididymis luminal protein 4; epididymis secretory protein Li 1; epididymis secretory protein Li 3; epididymis secretory protein Li 93; factor activating exoenzyme S; FAS; GW128; HEL4; HEL-S-1; HEL-S-3; HEL-S-93; HS1; KCIP-1; mitochondrial import stimulation factor S1 subunit; Msfs1; phospholipase A2; prepronerve growth factor RNH-1; Protein 1054; Protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; SEZ-2; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta polypeptide; | |
Mouse | |
Protein G | |
RUO | |
100153160, 22631, 25578, 286863, 287022, 475864, 477237, 54401, 56011, 7529, 7534, 780440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |