missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 eta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546811
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ADGNEKKLEK VKAYREKIEK ELETVCNDVL SLLDKFLIKN CNDFQYESKV Sequence homology: Dog: 100%; Human: 100%; Rabbit: 100%; Zebrafish: 78%
14-3-3 eta Polyclonal antibody specifically detects 14-3-3 eta in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
14-3-3 eta | |
Polyclonal | |
Unconjugated | |
YWHAH | |
14-3-3 eta; 14-3-3 protein eta; 14-3-3 protein eta-subtype; 14-3-3e; 1433F; fb93e02; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide; wu:fb51h04; wu:fb93e02; YWHA1; YWHA114-3-3 eta; ywhag; YWHAH; zgc:63883; zgc:63883 protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7533 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q04917 | |
YWHAH | |
Synthetic peptide directed towards the N-terminal of human YWHAH | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |