missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products 1422A DuPont™ Tyvek™ Autoclave Sterilization Wrapper
GREENER_CHOICEBIOPHARMA

Autoclavable Wrapper provides an excellent bacterial and microbial barrier for superior performance post steam sterilization.

Brand:  Keystone Cleanroom Products T2ASHEET0624

 View more versions of this product

Product Code. 17758527

  • £1200.00 / Pack of 500
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Tyvek™ Autoclavable Wrappers are breathable while providing an excellent bacterial and microbial barrier for superior performance post steam sterilization.

  • Lint-free (Helmke Drum) means no airborne particulates are generated during its use as sterilization packaging
  • Clean and consistently cut using an automated CAD system per ISO 8 Requirements
  • Packaged in an ISO 6 Controlled Clean Room
  • Designed for Steam Sterilization
  • Hydrophobic
  • Resists tearing and punctures
Specifications

Specifications

Sterilization Wrap
COC
Single-use
Pharmaceutical Production Equipment
Non-sterile
Yes
Steam, EtO, Gamma, VHP
<125°C
6 in.
24 in.
Autoclavable
White
Steam Sterilization
HDPE
5.3 mm
500
<250°F
6 x 24 in.
15.2 cm
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers