Learn More
Fisherbrand Vial screw neck with dropper assembly supplied not fitted to vial type 1 neutral borosilicate glass tube fitted to a PP cap with natural rubber bulb clear 47mm x 16mm (h x O.D.), 40µL dropper 4.5mL Fisherbrand
Brand:  Fisherbrand
Vial screw neck with dropper assembly supplied not fitted to vial type 1 neutral borosilicate glass tube fitted to a PP cap with natural rubber bulb clear 47mm x 16mm (h x O.D.), 40µL dropper 4.5mL Fisherbrand
Code : 33
Additional Details : Weight : 0.65000kg
Product Code. 14843572