Learn More
Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments
Ensure reliable connections for Thermo Scientific™ GC instruments with these Thermo Scientific™ Ferrules.
£48.26 - £124.00
Product Code | Brand | For Use With (Equipment) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Diameter (Metric) | Price | Quantity & Availability | ||||
10428874
|
Thermo Scientific™
29033460 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 0.32 mm |
£102.25
Pack of 10 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
||||
10478874
|
Thermo Scientific™
29033461 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 0.1 to 0.25 mm |
£102.25
Pack of 10 |
In Stock
Log in to see stock. |
||||
10660046
|
Thermo Scientific™
29033471 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 0.53 mm |
£105.32
Pack of 10 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
10466744
|
Thermo Scientific™
29033496 |
Thermo Scientific MS Detectors | 0.1 to 0.25 mm |
£74.46
Pack of 10 |
In Stock
Log in to see stock. |
||||
10571115
|
Thermo Scientific™
29033497 |
Thermo Scientific MS Detectors | 0.32 mm |
£124.00
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
10117644
|
Thermo Scientific™
290VP144 |
Agilent MS Detectors | 0.53 mm |
£48.26
Pack of 10 |
In Stock
Log in to see stock. |
||||
10595615
|
Thermo Scientific™
290VT165 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 6.35 mm |
£60.33
Pack of 10 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
10554125
|
Thermo Scientific™
290VT168 |
Agilent Injectors and Non-MS Detectors | 3.175 mm |
£59.30
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
10649485
|
Thermo Scientific™
290VT186 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 0.1 to 0.25 mm |
£55.15
Pack of 10 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
10630826
|
Thermo Scientific™
290VT187 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 0.32 mm |
£56.20
Pack of 10 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
10640826
|
Thermo Scientific™
290VT188 |
Thermo Scientific TRACE Ultra SSL Injectors and Non-MS Detectors | 0.53 mm |
£56.13
Pack of 10 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||||
Description
Use Thermo Scientific™ 15% Graphite/85% Vespel Ferrules for Thermo Scientific Instruments to ensure optimal performance of your Thermo Scientific GC instrument. They are available in three different materials and in various dimensions to accommodate a range of instruments and applications. All ferrules are are supplied in contaminant-free, individual blister packs.
FeaturesAvailable in 100% graphite, 15% graphite/85% vespel and SilTite metal
Ferrules for different types of injectors and detectors
Specifications
15% Graphite, 85% Vespel |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.