missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 150 Homogenizer Package
Includes everything necessary to process samples as small as 200μL to 100mL. Fisherbrand™ 150 Homogenizer Package includes the 150w motor as well as accessories such as 2 saw tooth stainless steel generator probes, disposable generator probes and more.
£4120.00
Specifications
Speed | 5000 to 35,000 rpm |
---|---|
Volume Range (Metric) | 200 μL to 100 mL |
Diameter (Metric) | 5.1 cm |
Height (Metric) | 16.5 cm |
For Use With (Application) | Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics |
Product Code | Brand | Plug Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Type | Price | Quantity & Availability | |||||
15595809
|
Fisherbrand™
|
UK Plug |
£4120.00
Each |
Estimated Shipment: 19-09-2024
Log in to see stock. |
|||||
Description
- Processes laboratory samples, from 200μL to 100mL
- Powerful 150w motor
- Lightweight ergonomic design
- Quiet operation, under 72db
- Available with disposable generator probes (adapter for use is included)
- Cross-contamination can be completely eliminated with the use of plastic generator probes
- Stainless steel generator probes are recommended for larger and tougher samples
Certifications
CE
Warranty
One year
Specifications
5000 to 35,000 rpm | |
5.1 cm | |
Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics | |
1 Ea. |
200 μL to 100 mL | |
16.5 cm | |
150 W motor, Flat bottom Stainless Steel generator probe (5 x 75 mm), Saw tooth Stainless Steel generator probe (7 x 115 mm), Saw tooth Stainless Steel generator probe (10 x 115 mm), Stand clamp, Motor stand assembly, Disposable probe adapter, Disposable generator probes, Instruction manual and Tool kit | |
466 g |