Learn More
3M™ 1500 Graffiti Remover
Removes graffiti, two types of remover depending on surface type
Brand:  3M™ 7000082039
158.87 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
Product Code. 17297610
  • £184.00 / Pack of 4
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has little odour, enhancing user comfort and efficiency – unlike traditional chemical removers which emit powerful odours and harmful vapours
  • Low environmental impact as the product does not contain aromatic, aliphatic or chlorinated solvents
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Graffiti Remover
500 mL