missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well Nontreated Cyclic Olefin Copolymer Plates
GREENER_CHOICE

Product Code. p-4681096 Shop All Corning Products
Click to view available options
Color:
Black, Clear Bottom
Quantity:
10/Pk.
20/Pk.
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 15512151

Brand: Corning™ 4565

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Nonsterile microplates have smooth top and clear, flat bottom.

Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.

  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
TRUSTED_SUSTAINABILITY

Specifications

Barcode Includes Barcode
Format Standard
Lid Without Lid
Culture Area 0.03 cm2
Array 32 x 48
Binding Type Not Treated
Color Black, Clear Bottom
Product Type 1536-Well Assay Microplate
For Use With (Equipment) Automated dispensers
Material Cycloolefin Copolymer
Volume (Metric) Well 12.5 μL
No. of Wells 1536
Volume (Metric) Working 8 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For higher sensitivity
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.