Learn More
Corning™ 1536-Well Nontreated Polystyrene Plates
GREENER_CHOICE
Superior flatness minimizes variability within a plate and between plates
Brand:  Corning™ 3725
 View more versions of this product
Product Code. 10465823
  • £1035.40 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Flat bottom wells are compatible with automated dispensers including the PerkinElmer™ FlexDrop™
  • Working volume of up to 10 μL
Generic Bar Code
  • Ensures compatibility with most bar code scanning and software systems
  • To avoid mix-ups, the surface treatment of the microplate wells is easily identified in the human readable portion of the bar code (NT = not treated; TC = tissue culture treated; NB = nonbinding surface)
  • Labels applied to all four sides of the plate No sequence number duplicates
Order Info
Packaged in bags of 10
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
32 x 48
Automated dispensers
1536
Non-sterile
Flat Bottom (F-well)
50/Cs.
Without Lid
White
Polystyrene
8 μL
Non-treated
For barcode scanning and software systems
10/Pk.