missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 1536-Well, Ultra-Low Binding, U-Shaped-Bottom Microplate
GREENER_CHOICE

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

Brand:  Corning™ 4527

Product Code. 15905981

  • £5545.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

With their novel and proprietary design, these microplates are ideal for generating and analyzing 3D multicellular spheroids in the same microplate. The Ultra-Low Attachment (ULA) surface enables uniform and reproducible 3D multicellular spheroid formation. The black opaque microplate body shields each optically clear, round bottom well from well-to-well cross-talk.

  • 1536-well format allows for high throughput 3D cell culture
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel well geometry aids in the generation of uniform, single spheroids across all wells, which enables automated visualization.
  • Unique design shields each well to minimize well-to-well cross-talk.
  • You can culture and assay spheroids in the same microplate, without the need for transfer to a new microplate.
  • Compatible with the GNF ultra high throughput system
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
32 x 48
Ultra-Low Attachment
Spheroid
14 μL
Sterile
Round Bottom (U-well)
50/Cs.
50/Cs.
125.75 mm
8 mm
With Lid
10/Pk., 50/Cs.
Black, Clear Bottom
Polystyrene
1536
Ultra-low Binding
3D Cell Culture
10/Pk.
Clear
84.12 mm
6.1 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers