Learn More
Corning™ 1536-Well White Round Bottom Polystyrene, Not Treated Microplate
White walled plates enhance luminescent signals and have low background luminescence and fluorescence
Brand: Corning™ 3937
This item is not returnable.
View return policy
Description
- Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
- Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
- Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm²
- Recommended working volume of up to 8 μL
- Conforms to standard microplate footprint and dimensions
Specifications
Without Lid | |
Polystyrene | |
Up to 8 μL | |
Non-treated | |
10/Pk., 50/Cs. |
White | |
1536 | |
Non-sterile | |
Round |