missing translation for 'onlineSavingsMsg'
Learn More

16-Hexadecanolide, 97%

Product Code. 11346686
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11346686

Brand: Thermo Scientific Alfa Aesar L03864.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-29-5 | C16H30O2 | 254.41 g/mol

16-Hexadecanolide is used in the manfacturing of perfumes. It acts as masking agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
16-Hexadecanolide is used in the manfacturing of perfumes. It acts as masking agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-29-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00039668
InChI Key LOKPJYNMYCVCRM-UHFFFAOYSA-N
Synonym 16-hexadecanolide, hexadecanolide, cyclohexadecanolide, 16-hexadecanolactone, 1,16-hexadecanolide, dihydroambrettolide, juniperic acid lactone, 1,16-hexadecanolactone, hexadecanolactone, 16-hydroxyhexadecanoic acid lactone
PubChem CID 7984
SMILES O=C1CCCCCCCCCCCCCCCO1

Specifications

Melting Point 33°C to 34°C
Flash Point >100°C (212°F)
Quantity 50 g
Beilstein 148631
Solubility Information Insoluble in water.
IUPAC Name oxacycloheptadecan-2-one
Formula Weight 254.42
Percent Purity 97%
Chemical Name or Material 16-Hexadecanolide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.