missing translation for 'onlineSavingsMsg'
Learn More

Tuttnauer 160L Benchtop Autoclave

The 5075EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, biohazard and waste sterilization, Fo control, and more.

Brand:  Tuttnauer 5075ELC-380-D


Additional Details : Weight : 203.00000kg

 View more versions of this product

Product Code. 17888988

  • £22522.88 / Each
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

Designed for your laboratory applications. The 5075EL-D has an advanced multi-color control panel and a chamber made of 316L stainless steel. Each model has a number of optional added-value features which can be configured for fast cooling, efficient drying, Fo control, and more.

  • Advanced Control System - Take Advantage of Tuttnauer’s state-of-the-art control system with multi-color display
  • Safe - Automatic pressure locking system. Door will not open when the chamber is overpressured
  • Optional Internal Printing - Documents date, time, temperature and pressure
  • Intuitive Multi-Color Display - Multi-color display for easier reading gives quick access to important information. Color is used to indicate the stage of the cycle
  • High Quality Stainless Steel - 316L grade stainless steel chamber and door with electro-polish finish
  • Diverse Load Types - The 5075EL-D autoclave comes with 8 factory set cycle programs that cover the processing needs of most laboratories and research centers
Specifications

Specifications

35.2 gal.
75 cm
30 in.
44.8 in.
Benchtop Autoclave
105°C to 137°C
316L Stainless Steel
160 L
20 in.
50 cm
112 cm
221°F to 279°F
Three Phase, Cooling
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers