missing translation for 'onlineSavingsMsg'
Learn More

Melford 17-AAG

Product Code. p-200086977
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
This item is not returnable. View return policy

Product Code. 30017965

Brand: Melford A200450.0005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

17-AAG (17-Allylamino-17-demethoxygeldanamycin) is a synthetic derivative of geldanamycin. Like geldanamycin, 17-AAG is recognized for its role as an inhibitor of heat shock protein 90 (Hsp90), a molecular chaperone involved in protein folding and stabilization.

17-AAG (17-Allylamino-17-demethoxygeldanamycin) is a synthetic derivative of geldanamycin. Like geldanamycin, 17-AAG is recognized for its role as an inhibitor of heat shock protein 90 (Hsp90), a molecular chaperone involved in protein folding and stabilization.

It is often used as a tool to study Hsp90-dependent processes. By inhibiting Hsp90, 17-AAG impacts the stability and function of various client proteins, many of which are involved in critical cellular processes, including signal transduction and cell cycle regulation. Similar to geldanamycin, 17-AAG is particularly relevant in cancer research, as Hsp90 is often crucial for the stability of oncoproteins.

Researchers use 17-AAG to explore the molecular mechanisms of Hsp90 inhibition, with potential applications in understanding cancer biology and developing therapeutic strategies.

Specifications

CAS 75747-14-7
Molecular Formula C31H43N3O8
Quantity 500 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.