missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ 1733 OSRAM Power Bank Battery Pack

A mobile, external battery with flash warning protection for devices that can be charged via USB ports.

Brand:  Uvex™ 1733200

13.87 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.



Additional Details : Weight : 0.09400kg Transport : UN number : 3480 Chem class : 9

Product Code. 17280301

  • £18.95 / Each
Estimated Shipment: 12-08-2024
to see stock.

Description

Description

  • Mobile, external battery for devices that are charged via USB
  • Rechargable with micro USB cable or charger
  • LED indicator
  • Delivery: Power Bank, micro-USB-wire
  • Capacity: 3000 mAh
Specifications

Specifications

Battery Pack
OSRAM POWERBANK 895065
Devices that are charged via USB
approx. 83 g
1 A
Yes
Lithium Polymer
LED Indicator
5 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers