Learn More
Testo™ 176 T2 Two-Channel Temperature Data Logger
Brand: Testo™ 0572 1762
Code : A0
Additional Details : Weight : 0.47700kg
Specifications
Lithium (TL-5903) | |
176 T2 Two-Channel Temperature Data Logger | |
4 x 2 x 1 in. (103 x 63 x 33 mm) |
8 years (15 min. meas. rate, +25°C) | |
2 External | |
103 mm |