missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Coverglass Forceps

Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Coverglass Forceps has 6mm wide points.

Brand:  Bochem™ 1722-10

 View more versions of this product

Product Code. 11885121

This item is not returnable. View return policy

Description

Description

  • Coverglas forceps
  • Stainless steel
  • Points 6mm wide
Specifications

Specifications

Bent
105 mm
1 Pack
Coverglass Forceps
Stainless Steel (18/10)
Forceps
Good grip risk-free membranes, autoclaving or flame, pliers
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers