missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Double Spatula, Round Bent

Use the Bochem™ 18/10 Stainless Steel Double Spatula, Round Bent for your lab needs.

Brand:  Bochem™ 3187

 View more versions of this product

Product Code. 10234131

  • £4.15 / Each
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless
  • Rounded Edge
Specifications

Specifications

1 Ea.
Spoon End, Bent End
25 mm
12 mm
4mm
Stainless Steel (18/10)
Autoclavable
Double Ended Spatula
12 mm
Stainless Steel (18/10)
210 x 12 mm
210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers