missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Double Spatula, Round Bent
Use the Bochem™ 18/10 Stainless Steel Double Spatula, Round Bent for your lab needs.
Brand: Bochem™ 3184
This item is not returnable.
View return policy
Description
- Stainless
- Rounded Edge
Specifications
1 Ea. | |
Spoon End, Round End | |
17 mm | |
7 mm | |
2.5mm | |
Stainless Steel (18/10) |
Autoclavable | |
Double Ended Spatula | |
7 mm | |
Stainless Steel (18/10) | |
200 x 7 mm | |
200 mm |