missing translation for 'onlineSavingsMsg'
Learn More

Agencinox™ 18/10 Stainless Steel High Evaporating Dish

Use these dishes for general evaporation applications. Agencinox™ 18/10 Stainless Steel High Evaporating Dishes are ideal for use in medical environments.

Brand:  Agencinox™ 1140

Additional Details : Weight : 0.26000kg

 View more versions of this product

Product Code. 11731923

  • £57.65 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • High form evaporating dish
  • Stainless steel construction
  • Available in a variety of sizes
Specifications

Specifications

Flat
Stainless Steel
Evaporating Dish
70 mm
1 L
Round
Evaporating Application
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers