missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Laboratory Bowl
Store, mix or contain chemicals with the Bochem™18/10 Stainless Steel Laboratory Bowl. This is a useful laboratory accessory.
Brand: Bochem™ 8597
This item is not returnable.
View return policy
Description
- Flat bottom
- Stainless steel
- Spherical bowl
Specifications
Flat | |
Stainless Steel | |
Laboratory Bowl | |
400 mm |
14 L | |
Round | |
160 mm |