Learn More
Bochem™ 18/10 Stainless Steel Standard Spoon Spatula
Manufactured from 18/10 Stainless Steel. BOCHEM™ 18/10 Stainless Steel Standard Spoon Spatula is a standard spatula.
Brand: Bochem™ 3215
Description
- Stainless steel
Specifications
1 Ea. | |
Spoon Spatula | |
4mm | |
230 mm |
Autoclavable | |
Designed for a wide range of uses | |
Stainless Steel (18/10) |