missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ 18/10 Stainless Steel Transport jug
£994.00 - £1280.00
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10642692
|
Bochem™
8381 |
220 mm |
£994.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
10594261
|
Bochem™
8384 |
270 mm |
£1280.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Specifications
Stainless Steel | |
Cylindrical | |
Transport Jug |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title