missing translation for 'onlineSavingsMsg'
Learn More

Motic™ 18/28 Series Cordless LED Microscope

Ideally suited microscope for academic microscopy. Motic™ 18/28 Series Cordless LED Microscope offers a unique ergonomic shape, superior optics and portability.

Brand:  Motic™ 1100102201044

 View more versions of this product

Product Code. 15495102

  • £633.00 / Each
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Bright white LED illumination with no heat.
  • Rechargeable battery pack facilitates cordless use.
  • Monocular, binocular and trinocular versions available.
  • DIN EA Achromatic objectives 4X, 10X and 40X
  • Separated and coaxial coarse and fine focusing with a minimum increment of 2μm to ensure a perfectly focused sample.
  • Integrated mechanical stage available.
Specifications

Specifications

Rack and pinion mounted 1.25 N.A. Abbe condenser with iris diaphragm and filter holder
WF 10X/18 mm
LED
2823 LED Cordless Microscope
Cordless LED Microscope
Reverse-mounted, quadruple revolving nosepiece with positive click stops for precise magnification changes
Separate coarse and fine focusing system with upper limit stop to prevent damage to slides and objectives and focus tension adjustment to prevent stage drift.
Trinocular with diopter adjustment, 30° inclined, 360° rotating, light split 50/50
4X, 10X, 40XS
EA Achromatic, Parfocal and Parcentric
Large 133mm x 136mm mechanical stage with low-position coaxial controls
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.