missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter for 18-place Rotor
Designed for use with Eppendorf Cryovials™ and HPLC Tubes
Brand: Eppendorf™ 5427708006
124.00 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Adapter
- For 1 HPLC vessel 1.5 mL, for Rotor F-45-18-17-Cryo
Specifications
Adapter | |
1.5mL HPLC Vials | |
18/Pk. |