missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-(2-naphthyloxy)propanoic acid, 97% 1g

Brand:  Thermo Scientific Maybridge JM00276DA

2-Methyl-2-(2-naphthyloxy)propanoic acid, 97% 1g

Code : 31

Additional Details : CAS Number : 7756-78-7 Weight : 0.05100kg

Product Code. 10015522

  • £21.95 / 1g
Estimated Shipment: 12-11-2024
to see stock.