missing translation for 'onlineSavingsMsg'
Learn More
1H-1,2,3-Triazole, 98%, Thermo Scientific Chemicals Product Code.: 11438846

1H-1,2,3-Triazole, 98%, Thermo Scientific Chemicals

Product Code. 11438846
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11438846

Brand: Thermo Scientific Alfa Aesar A16999.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It effectively promotes the proton conduction in polymer electrolyte membranes via an intermolecular proton-transfer mechanism. A useful triazole for proteomics research

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-36-8
Molecular Formula C2H3N3
Molecular Weight (g/mol) 69.067
MDL Number MFCD00014490
InChI Key QWENRTYMTSOGBR-UHFFFAOYSA-N
Synonym 1h-1,2,3-triazole, 1,2,3-triazole, 2h-1,2,3-triazole, 1,2,3-1h-triazole, triazole, triazol, osotriazole, v-triazole, 1,2,3-triazol, 1,2,3 triazoleShow MoreShow Less
PubChem CID 67516
ChEBI CHEBI:35565
IUPAC Name 2H-triazole
SMILES C1=NNN=C1

Specifications

Melting Point 23°C to 25°C
Density 1.192
Boiling Point 208°C
Flash Point 107°C (224°F)
Refractive Index 1.498
Quantity 1 g
Beilstein 104766
Solubility Information Fully miscible in water.
Formula Weight 69.07
Percent Purity 98%
Chemical Name or Material 1H-1,2,3-Triazole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10205121 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16999.03,A16999.14,A16999.06
Lot NumberE7725B Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16999.03,A16999.14,A16999.06
Lot Number10226517 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16999.03,A16999.14,A16999.06
Lot Number10100233 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16999.03,A16999.14,A16999.06
Lot Number10223819 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16999.03,A16999.14,A16999.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-1,2,3-Triazole, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.