missing translation for 'onlineSavingsMsg'
Learn More
1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%, Thermo Scientific™ Product Code.: 11399255

1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%, Thermo Scientific™

Product Code. 11399255
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11399255

Brand: Thermo Scientific Alfa Aesar L16589.03

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H,2H-Perfluoro-1-dodecanol is used in fabric processing, coating, appliance paint and boat paint.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-86-1
Molecular Formula C12H5F21O
Molecular Weight (g/mol) 564.14
MDL Number MFCD00039545
InChI Key FLXYIZWPNQYPIT-UHFFFAOYSA-N
Synonym 2-perfluorodecyl ethanol, 1h,1h,2h,2h-perfluorododecan-1-ol, 1,1,2,2-tetrahydroperfluoro dodecanol, 1h,1h,2h,2h-perfluoro-1-dodecanol, unii-a72u7q973t, 1h,1h,2h,2h-perfluorododecanol, 1,1,2,2-tetrahydroperfluoro-1-dodecanol, 1h,1h,2h,2h-perfluorodeca-1-ol, 1h,1h,2h,2h-perfluoro-1-dodecanol 1g
PubChem CID 70083
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododecan-1-ol
SMILES OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point 90°C to 95°C
Boiling Point 145°C (10 mmHg)
Linear Formula CF3(CF2)9CH2CH2OH
Quantity 1 g
Beilstein 2029867
Solubility Information Insoluble in water.
Formula Weight 564.14
Percent Purity 96%
Chemical Name or Material 1H,1H,2H,2H-Perfluoro-1-dodecanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H,1H,2H,2H-Perfluoro-1-dodecanol, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.