missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorodecyl acrylate, 96%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific™

Product Code. 15440827
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27905-45-9
Melting Point -3°C
Density 1.637
Boiling Point 89°C to 90°C (4 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C13H7F17O2
Refractive Index 1.337
MDL Number MFCD00042306
Quantity 5 g
Beilstein 5173337
Synonym 1h,1h,2h,2h-heptadecafluorodecyl acrylate, 1h,1h,2h,2h-perfluorodecyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl acrylate, unii-y9bfb4689z, 2-perfluorooctyl ethyl acrylate, 1,1,2,2-tetrahydroperfluorodecyl acrylate, perfluorooctyl ethyl acrylate, 2-propenoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl ester, 2-perfluorooctyl ethylacrylate, c8f17ch2ch2ococh=ch2
InChI Key QUKRIOLKOHUUBM-UHFFFAOYSA-N
SMILES C=CC(=O)OCCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl prop-2-enoate
Molecular Weight (g/mol) 518.17
PubChem CID 119747
Formula Weight 518.17
Percent Purity 96%
Chemical Name or Material 1H,1H,2H,2H-Perfluorodecyl acrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'