missing translation for 'onlineSavingsMsg'
Learn More
1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%, Thermo Scientific Chemicals Product Code.: 11339285

1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11339285
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11339285

Brand: Thermo Scientific Alfa Aesar L16606.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H,1H,2H,2H-Perfluorooctyltrichlorosilane is used in the silanization of silicon wafer. Self assembled monolayers treated with this, acts as an anti adhesive layer and release of cured poly(dimethyl siloxanes). It is also used to fabricate chemical surface patterns of self assembled monolayers.

Solubility
Miscible with tetrahydrofuran, tetrhydropyran, toluene and other organic solvents.

Notes
Incompatible with water, strong oxidizing agents, alcohols, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78560-45-9
Molecular Formula C8H4Cl3F13Si
Molecular Weight (g/mol) 481.534
MDL Number MFCD00042363
InChI Key PISDRBMXQBSCIP-UHFFFAOYSA-N
Synonym trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, 1h,1h,2h,2h-perfluorooctyltrichlorosilane, silane, trichloro 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, tridecafluoro-1,1,2,2-tetrahydrooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluorooctyl silane, 2-tridecafluorohexyl ethyltrichlorosilane, 1h,1h,2h,2h-perfluorooctyl trichlorosilane, trichloro 1h,1h,2h,2h-perfluoro-n-octyl silane, trichloro 1h,1h,2h,2h-tridecafluoro-n-octyl silane
PubChem CID 123578
IUPAC Name trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C(C[Si](Cl)(Cl)Cl)C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Boiling Point 192°C
Quantity 50 g
UN Number UN2987
Beilstein 6154526
Flash Point 87°C (189°F)
Solubility Information Miscible with tetrahydrofuran,tetrhydropyran,toluene and other organic solvents.
Formula Weight 481.55
Percent Purity 97%
Refractive Index 1.352
Sensitivity Moisture sensitive
Density 1.638
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H,1H,2H,2H-Perfluorooctyltrichlorosilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.