missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzotriazole-(ring-d4) solution, Honeywell Fluka™

Product Code. 15949681 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 15949681

Brand: Honeywell Fluka R11645ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

10 μg/mL in acetone (15949681), 50 μg/mL in acetone (15900192) Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 67-64-1
Density 0.79 g/cm3
Flash Point -17°C
Physical Form Liquid
Boiling Point 55°C to 57°C
Industry Type Parabenes
Quantity 5 mL
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.