missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Indazole-4-carboxylic acid, 97%, Thermo Scientific™
£137.00 - £345.36
Chemical Identifiers
CAS | 677306-38-6 |
---|---|
Molecular Formula | C8H6N2O2 |
Molecular Weight (g/mol) | 162.15 |
MDL Number | MFCD03840644 |
InChI Key | KGKZHHIUOZGUNP-UHFFFAOYSA-N |
Synonym | indazole-4-carboxylic acid, 1h-indazole-4-carboxylicacid, 4-indazolecarboxylic acid, 4-carboxy-1h-indazole, 2h-indazole-4-carboxylic acid, pubchem19651, acmc-1b3os, ksc352q1d, 1h-indazole-4-carboxylic acid, 4-1h indazole carboxylic acid |
PubChem CID | 21982323 |
IUPAC Name | 1H-indazole-4-carboxylic acid |
SMILES | C1=CC(=C2C=NNC2=C1)C(=O)O |
Description
Chemical Identifiers
677306-38-6 | |
162.15 | |
KGKZHHIUOZGUNP-UHFFFAOYSA-N | |
21982323 | |
C1=CC(=C2C=NNC2=C1)C(=O)O |
C8H6N2O2 | |
MFCD03840644 | |
indazole-4-carboxylic acid, 1h-indazole-4-carboxylicacid, 4-indazolecarboxylic acid, 4-carboxy-1h-indazole, 2h-indazole-4-carboxylic acid, pubchem19651, acmc-1b3os, ksc352q1d, 1h-indazole-4-carboxylic acid, 4-1h indazole carboxylic acid | |
1H-indazole-4-carboxylic acid |
Specifications
677306-38-6 | |
100.0 | |
White to Cream | |
96% min. (HPLC) | |
C8H6N2O2 | |
indazole-4-carboxylic acid, 1h-indazole-4-carboxylicacid, 4-indazolecarboxylic acid, 4-carboxy-1h-indazole, 2h-indazole-4-carboxylic acid, pubchem19651, acmc-1b3os, ksc352q1d, 1h-indazole-4-carboxylic acid, 4-1h indazole carboxylic acid | |
C1=CC(=C2C=NNC2=C1)C(=O)O | |
162.15 | |
162.15 | |
Crystalline Powder |
96.0 | |
209.5°C to 211.0°C | |
Authentic | |
Glass Bottle | |
MFCD03840644 | |
KGKZHHIUOZGUNP-UHFFFAOYSA-N | |
1H-indazole-4-carboxylic acid | |
21982323 | |
97% | |
1H-Indazole-4-carboxylic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1H-Indazole-4-carboxylic acid, 97%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only