missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-6-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11966641
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 669050-69-5
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
InChI Key JTWYTTXTJFDYAG-UHFFFAOYSA-N
Synonym 1h-indazole-6-carboxaldehyde, 6-formylindazole, 6-formyl-1h-indazole, 2h-indazole-6-carbaldehyde, indazole-6-carboxaldehyde, 1h-indazole-6-cabaldehyde, pubchem23878, acmc-1bc45, 2h-indazole-6-carboxaldehyde, 1-h-indazole-6-carboxaldehyde
PubChem CID 18538368
IUPAC Name 1H-indazole-6-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)NN=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180.6°C to 180.8°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 5 g
Formula Weight 146.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1H-Indazole-6-carboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazole-6-carboxaldehyde, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'