missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-2-methanol, 96%, Thermo Scientific Chemicals

Product Code. 15592373
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24621-70-3
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.17
InChI Key XEEANGGQJOWRTG-UHFFFAOYSA-N
Synonym 1h-indol-2-yl methanol, 1h-indole-2-methanol, 1h-indol-2-yl-methanol, 2-hydroxymethylindole, 2-hydroxymethyl indole, hydroxymethylindol, 2-indolemethanol, indole-2-methanol, indol-2-ylmethan-1-ol
PubChem CID 98783
IUPAC Name 1H-indol-2-ylmethanol
SMILES C1=CC=C2C(=C1)C=C(N2)CO

Specifications

Melting Point 72.0°C to 78.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Quantity 5 g
Formula Weight 147.17
Percent Purity 96%
Chemical Name or Material 1H-Indole-2-methanol
compliance-icons
Product Identifier
  • 1H-Indole-2-methanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'