Learn More
1H-Indole-3-acetic acid, 98%, Thermo Scientific Chemicals
1H-Indole-3-acetic acid, CAS # 87-51-4, also known as indole-3-acetic acid, is a monocarboxylic acid mainly used as a plant growth hormone and is the most common naturally occurring plant hormone of the auxin family.
£30.75 - £598.98
Chemical Identifiers
CAS | 87-51-4 |
---|---|
Molecular Formula | C10H9NO2 |
Molecular Weight (g/mol) | 175.19 |
MDL Number | MFCD00005636 |
InChI Key | SEOVTRFCIGRIMH-UHFFFAOYSA-N |
Synonym | indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a |
PubChem CID | 802 |
ChEBI | CHEBI:16411 |
IUPAC Name | 2-(1H-indol-3-yl)acetic acid |
SMILES | OC(=O)CC1=CNC2=CC=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10275350
|
Thermo Scientific Acros
122160100 |
10 g | Glass bottle |
£30.75
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10449040
|
Thermo Scientific Acros
122160250 |
25 g | Glass bottle |
£51.56
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285350
|
Thermo Scientific Acros
122161000 |
100 g | Glass bottle |
£163.82
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10043613
|
Thermo Scientific Acros
122165000 |
500 g | Glass bottle |
£598.98
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• 1H-Indole-3-acetic acid is a plant growth hormone that belongs to the auxin class. It is also considered to be an inducer of plant cell elongation and division that can cause uncontrolled growth
Applications
• This compound is a monocarboxylic acid; an acetic acid in which one of the methyl hydrogens has been replaced by a 1H-indol-3-yl group.
• Beyond its role as a plant hormone, this compound regulates tissue differentiation, apical dominance, and responses to light, gravity, and pathogens
• It can be used as a signaling molecule necessary for development of plant organs and coordination of growth
Chemical Identifiers
87-51-4 | |
175.19 | |
SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
802 | |
2-(1H-indol-3-yl)acetic acid |
C10H9NO2 | |
MFCD00005636 | |
indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a | |
CHEBI:16411 | |
OC(=O)CC1=CNC2=CC=CC=C12 |
Specifications
87-51-4 | |
100.0 | |
Colorless to Yellow | |
Authentic | |
C10H9NO2 | |
22, 66 | |
indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a | |
SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
2-(1H-indol-3-yl)acetic acid | |
802 | |
175.19 | |
Liquid |
99.0 | |
167.0°C | |
171°C | |
98% (HPLC) | |
MFCD00005636 | |
15, 5004 | |
Solubility in water: 8g/L (20°C). Other solubilities: soluble in acetone and ether,freely soluble in alcohol,sparingly soluble in chloroform | |
OC(=O)CC1=CNC2=CC=CC=C12 | |
175.19 | |
CHEBI:16411 | |
98% | |
1H-Indole-3-acetic acid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-748-2
- 2-(1H-indol-3-yl)acetic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only