missing translation for 'onlineSavingsMsg'
Learn More
1H-Pyrazole-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals Product Code.: 11395465

1H-Pyrazole-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Product Code. 11395465
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11395465

Brand: Thermo Scientific Alfa Aesar L19654.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1H-Pyrazole-4-boronic acid pinacol ester is a useful reagent for Suzuki-Miyaura cross-couplings as well as Ruthenium-catalyzed asymmetric hydrogenation. It is also used as reagent for preparing VEGF, Aurora, RHO (ROCK), Janus Kinase 2, c-MET, ALK, S-nitrsoflutathione reductase, CDC7, Acetyl-CoA carboxylase inhibitors as well as other biologically active compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-4-boronic acid pinacol ester is a useful reagent for Suzuki-Miyaura cross-couplings as well as Ruthenium-catalyzed asymmetric hydrogenation. It is also used as reagent for preparing VEGF, Aurora, RHO (ROCK), Janus Kinase 2, c-MET, ALK, S-nitrsoflutathione reductase, CDC7, Acetyl-CoA carboxylase inhibitors as well as other biologically active compounds.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 269410-08-4
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.04
MDL Number MFCD03453063
InChI Key TVOJIBGZFYMWDT-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, pyrazole-4-boronic acid pinacol ester, 4-pyrazoleboronic acid pinacol ester, 1h-pyrazole-4-boronic acid pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-4-boronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-1h-pyrazol-4-yl-1,3,2-dioxaborolane, pyrazole-4-boronic acid, pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 2-4-pyrazolyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2774010
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CC1(C)OB(OC1(C)C)C1=CNN=C1

Specifications

Melting Point 145°C to 149°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 194.04
Percent Purity 98%
Chemical Name or Material 1H-Pyrazole-4-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-Pyrazole-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.